missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (UM-A9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34507UV
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| DyLight 350 | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Mouse | |
| 205 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3691 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| UM-A9 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| ITGB4 | |
| Human squamous cell carcinoma (UM-SCC1) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 205kDa, which is identified as integrin beta-4 (ITGB4). Its epitope is localized in the extracellular domain of ITGB4 protein. Integrins are heterodimers comprised of alpha and beta subunits, that are non-covalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta-4 subunit, a receptor for the laminins. This subunit tends to associate with alpha-6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction