missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 6 Antibody (268622), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2389X
This item is not returnable.
View return policy
Description
Integrin beta 6 Monoclonal antibody specifically detects Integrin beta 6 in Mouse samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| Integrin beta 6 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 268622 | |
| Western Blot, Immunocytochemistry | |
| integrin beta-6, integrin, beta 6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6, Gly22-Asn706, Accession # Q9Z0T9 | |
| 0.1 mL | |
| Cancer | |
| 3694 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction