missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 6 Antibody (OTI1D2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71051AF647
This item is not returnable.
View return policy
Description
Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Integrin beta 6 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ITGB6 | |
| Full length recombinant protein of human Integrin beta 6 (NP_000879) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1D2 | |
| Western Blot | |
| integrin beta-6, integrin, beta 6 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 3694.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction