missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (2G18), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-94147AF488
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ITGB7 | |
| Cell lysates of mouse lymphocytes | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2G18 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction