missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (2G18), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-94147UV
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| DyLight 350 | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2G18 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ITGB7 | |
| Cell lysates of mouse lymphocytes | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction