missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (332418) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3060JF669
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin β7, Glu20-His722, Accession # P26011 | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 332418 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction