missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (473207), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4669I
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 473207 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform, Glu20-His723, Accession # P26010 | |
| 0.1 mL | |
| Signal Transduction | |
| 3695 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction