missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 8 Antibody (2723C), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB47752E
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| Integrin beta 8 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 2723C | |
| Flow Cytometry, Immunocytochemistry | |
| integrin beta-8, integrin, beta 8 | |
| Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 8, Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 8 (Glu43-Arg684), Accession # NP_002201.1 | |
| 0.1 mL | |
| Signal Transduction | |
| 3696 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction