missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 8 Antibody (416922) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4775JF525
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Integrin beta 8 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| integrin beta-8, integrin, beta 8 | |
| Mouse myeloma cell line NS0-derived recombinant human Integrin β8, Glu43-Arg684 (predicted), Accession # P26012 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 416922 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Signal Transduction | |
| 3696 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction