Learn More
Integrin αE2 (CD103) Mouse anti-Rat, BV605, Clone: OX-62, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 745138
This item is not returnable.
View return policy
Description
Integrin αE2 (CD103) Monoclonal antibody specifically detects Integrin αE2 (CD103) in Rat samples. It is validated for Flow Cytometry.
Specifications
| Integrin αE2 (CD103) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-62 | |
| Brilliant Violet 605 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |