missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intelectin-1/Omentin Mouse anti-Human, Clone: ITLN1/4064, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07689-100ug
This item is not returnable.
View return policy
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Intelectin-1/Omentin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin | |
| Recombinant fragment (around aa16-141) of human Intelectin-1/Omentin protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Peptide Array | |
| ITLN1/4064 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55600 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction