missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon alpha-21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597860
This item is not returnable.
View return policy
Description
Interferon alpha-21 Polyclonal Antibody for Western Blot, IP, ELISA
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Interferon alpha-21 | |
| Polyclonal | |
| Unconjugated | |
| IFNA21 | |
| IFNA21; IFNA5; IFNA6; IFN-alpha-21; IFN-alpha-5; IFN-alpha-6; IFN-alphaG; IFN-alphaI; IFN-alphaK; INA5; INFA5; Interferon; interferon alpha 21; interferon alpha 5; interferon alpha 6; interferon alpha-21; Interferon alpha-5; interferon alpha-54; Interferon alpha-6; interferon alpha-61; Interferon alpha-F; interferon alpha-G; Interferon alpha-K; interferon, alpha 21; interferon, alpha 5; interferon, alpha 6; LeIF F; LeIF G; LeIF K; leIF-F; leukocyte interferon protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 3452 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 3.4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P01568 | |
| IFNA21 | |
| Recombinant Human Interferon alpha-21 protein (24-189AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction