Learn More
B Braun™ Intrafix™ SafeSet Premium IV Administration Set
B Braun™ Intrafix™ SafeSet Premium IV Administration Set is a highly sophisticated, vented IV administration for gravity and pressure infusion.
Brand: B Braun™ 4063000
Additional Details : Weight : 1.50000kg
Description
- Suitable for gravity infusion and pressure infusion with defined, non-dedicated infusion pumps (pressure resistant up to 2 bar).
- Ergonomically designed 3-part drip chamber for comfortable, secure handling.
- Advanced precise roller clamp.
- Soft, clear PVC tubing with new type of safe plasticizer.
- AirStop: an innovative, hydrophilic depth filter membrane ensuring AirStop safety in case of emptied fluid container. Maintains the tubing completely filled with fluid. Works further as a particle filter with a filtration efficiency of 15μm and even lower.
- PrimeStop: a protective cap lined with a bacteria-tight hydrophobic membrane placed on the Luer Lock fitting, stops fluid leaking and protects against contamination. PrimeStop automates priming and ends messy dripping.
- Intrafix SafeSet helps prevent air infusion and infusion-related infection, reduces work load, saves time and costs and ensures a closed system during priming.
Specifications
ISO 8534-4/8 | |
180 cm | |
2 bar |
AirStop, PrimeStop | |
Vented IV administration set for gravity or pressure infusion |