Learn More
B Braun™ FEP Safety IV Catheters, Introcan
Product Code. 12301113
Change view
Click to view available options
Needle Gauge:
18 G
22 G
24 G
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
12301113 18 G Pack of 50
10453202 24 G Pack of 50
10174713 22 G Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12301113 Supplier B Braun™ Supplier No. 425456201
Use B Braun™ FEP Safety IV Catheters, Introcan™. From insertion to catheter deployment to needle removal, protection is provided by an automatically activated system that cannot be bypassed.
  • Color-coded labels for easy gauge size identification
  • Back cut bevel
  • FEP for firmer construction for arterial access and special procedures
  • DEHP- and latex-free
Specifications
Needle Gauge 18 G
Length (Metric) 32 mm
Product Type Introcan