missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-509G
This item is not returnable.
View return policy
Description
INTS1 Polyclonal antibody specifically detects INTS1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| INTS1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp586J0619, FLJ46624, integrator complex subunit 1, NET28, UNQ1821/PRO3434 | |
| A synthetic peptide derived from a sequence between residues 150 and 200 of human KIAA1440 using the numbering given in entry XP_291222.2 (GeneID 26173). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 26173 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction