missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INTS10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562216
This item is not returnable.
View return policy
Description
Immunogen sequence: RGENLFLKAV NKICQQGNFQ YENFFNYVTN IDMLEEFAYL RTQEGGKIHL ELLPNQGMLI KHHTVTRGIT KGVKEDFRLA MERQVSRCGE NLMVVLHRFC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1.
Specifications
| INTS10 | |
| Polyclonal | |
| Unconjugated | |
| Ints10 | |
| 4921521J11Rik; AI462004; C8orf35; int10; Integrator complex subunit 10; INTS10; RGD1304711 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55174 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVR2 | |
| Ints10 | |
| Recombinant protein corresponding to Human INTS10. Recombinant protein control fragment (Product #RP-103080). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction