missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS5 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60461H
This item is not returnable.
View return policy
Description
INTS5 Polyclonal antibody specifically detects INTS5 in Human samples. It is validated for Western Blot
Specifications
| INTS5 | |
| Polyclonal | |
| Western Blot | |
| Int5, integrator complex subunit 5, KIAA1698INT5 | |
| The immunogen recognized by this antibody maps to a region between residue 975 and the C-terminus of human integrator complex subunit 5 using the numbering given in entry NP_085131.1 (GeneID 80789). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80789 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction