missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTU Polyclonal Antibody
GREENER_CHOICE

Product Code. 15715681 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15715681 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15715681 Supplier Invitrogen™ Supplier No. PA557785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NDNGPVSILK HQSNQKTGVI VQQRYKDVNV YVNPKKLTVI KAKEQLKLLE VLVGIIHQTK WSWRRTGKQG DGERLVVHGL LPGGSAMKSG QVLIGDV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

Plays a key role in ciliogenesis and embryonic development. Regulator of cilia formation by controlling the organization of the apical actin cytoskeleton and the positioning of the basal bodies at the apical cell surface, which in turn is essential for the normal orientation of elongating ciliary microtubules. Plays a key role in definition of cell polarity via its role in ciliogenesis but not via conversion extension. Has an indirect effect on hedgehog signaling. Proposed to function as core component of the CPLANE (ciliogenesis and planar polarity effectors) complex involved in the recruitment of peripheral IFT-A proteins to basal bodies (PubMed:27158779). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTU
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTU
Gene Accession No. Q9ULD6
Gene Alias 9230116I04Rik; 9430087H23Rik; homolog of inturned; INT; Intu; Inturned planar cell polarity effector homolog; inturned planar cell polarity effector homolog (Drosophila); inturned planar cell polarity protein; KIAA1284; mKIAA1284; PDZ domain containing 6; PDZ domain-containing protein 6; PDZD6; PDZK6; Protein inturned
Gene Symbols INTU
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INTU. Recombinant protein control fragment (Product #RP-103774).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.