Learn More
Invitrogen™ ChargeSwitch™ iPrep™ iPrep ChargeSwitch Forensic Kit
Simple, three-step procedure in aqueous buffers
Merk:  Invitrogen™ IS10002
Overige gegevens : Gewicht : 1.91000kg
Produktcode. 10432123
  • € 375.00 / 1 kit
Geschatte verzenddatum
bij leverancier 29-05-2019
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Designed specifically for use with the iPrep Purification Instrument, the iPrep ChargeSwitch Forensic Kit takes advantage of the powerful ChargeSwitch technology, a major advance in DNA purification. This technology is based on a unique, ionizable nucleic acid-binding ligand whose charge can be switched based on the pH of the surrounding medium. Purification procedure avoids the use of guanidine, ethanol, and other troublesome reagents.
  • Flexibility in starting sample type, for quick and easy processing of different samples simultaneously
  • High sensitivity with samples containing very low amounts of DNA
  • Fast automated extraction time of 20 minutes
  • Standardized procedures minimize human error; preset protocols and prefilled cartridges eliminate variation
  • Offers single protocol for efficiently isolating DNA from all kinds of forensic samples, even if the DNA copy number is Low or DNA is partially degraded
  • Contents and Storage: iPrep™ ChargeSwitch Forensic Kit provides all reagents (supplied in prefilled cartridges), tubes, and tips needed for isolating DNA from forensic cell samples
  • Reagent cartridges can be stored at room temperature
  • Store lysis buffer and proteinase K at 4°C
Automated Genomic DNA Purification, Automated Nucleic Acid Purification, Clinical, DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, Sample Prep/Extraction for Human Identification, gDNA from Forensic Samples
Specificatie
Specificatie
Cloning, Forensic Testing, PCR, Sequencing, Southern Blotting
52 preps
Forensic DNA
ChargeSwitch™ Chemistry
Forensic Samples
Video's
For Research Use Only. Not for use in diagnostic procedures.