Learn More
Invitrogen™ 50 mL Tube Magnetic Separator
Product Code. 10228012
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10228012 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10228012 Supplier Invitrogen™ Supplier No. CS15050
Includes
One Magnetic Separator
Provides effective magnetic strength from neodymium magnets, which are aligned with plate wells or tubes.
  • Compatible with volumes used in kit protocols
  • Provides effective magnetic strength from neodymium magnets, which are aligned with plate wells or tubes
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Content And Storage Contains one Magnetic Separator, which may be stored at room temperature.
For Use With (Equipment) MagnaRack™ Magnetic Separator, Magna-Sep™ Magnetic Particle Separator
Shipping Condition Room Temperature
Product Line MagnaRack
Quantity 1 Each
Type Magnetic Separator