Learn More
Invitrogen™ Zero Background™ /Kan Cloning Kit
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Includes
25μg supercoiled pZErO-1 (the Zero Background Cloning Kit)or pZErO-2 vector (the Zero Background/Kan Cloning Kit) in TE, 200ng blunt-end test inserts, TOP10 E. coli stab, 25μL T4 DNA ligase, 10X ligation buffer, 1mL sterile water, and 2mL sterile TE. The Zero Background Cloning Kit also contains 1.25mL Zeocin selection agent.
- pZErO vectors use positive selection to eliminate high background
- pZErO-1 vector takes advantage of small size of Zeocin resistance gene to reduce the overall vector size for increased ligation and transformation efficiencies
- pZErO-2 vector includes the kanamycin resistance gene
Cloning, PCR Cloning
Specifications
Specifications
| Bacterial or Yeast Strain | TOP10 |
| Content And Storage | Store TOP10 stab at room temperature. Store the remaining kit components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Quantity | 25 μg |
| Vector | TA Cloning Vectors, pCR |