missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Antibody (IVRN/827), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11480MFV610
This item is not returnable.
View return policy
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human samples. It is validated for Flow Cytometry
Specifications
| Involucrin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| IVRN/827 | |
| Flow Cytometry | |
| involucrin | |
| Purified involucrin from human keratinocytes (Uniprot: P07476) | |
| 0.1 mL | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction