missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Antibody (rIVRN/9323), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24321AF488
This item is not returnable.
View return policy
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Involucrin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| involucrin | |
| Recombinant human full-length Involucrin protein (IVRN) | |
| 0.1 mL | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rIVRN/9323 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction