missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Antibody (SY5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2727SS
This item is not returnable.
View return policy
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Involucrin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 0.5 - 1.0 ug/10^6 cells, Immunohistochemistry 0.1-0.2 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
| involucrin | |
| Mouse | |
| 118 kDa | |
| 0.025 mg | |
| Primary | |
| Human, Pig, Canine, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| SY5 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| IVL | |
| Purified involucrin from human keratinocytes. | |
| Protein A purified | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction