missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Antibody (SY8), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2774R
This item is not returnable.
View return policy
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Involucrin | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IVL | |
| Purified involucrin from human keratinocytes. | |
| 0.1 mL | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SY8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| involucrin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Pig, Canine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction