missing translation for 'onlineSavingsMsg'
Learn More

Involucrin Antibody (SY8), FITC, Novus Biologicals™

Product Code. 18001384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001384 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001384 Supplier Novus Biologicals Supplier No. NB1002774F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Involucrin
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SY8
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias involucrin
Gene Symbols IVL
Host Species Mouse
Immunogen Purified involucrin from human keratinocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Human, Pig, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.