missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Mouse, Clone: SPM259, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34354-0.02MG
This item is not returnable.
View return policy
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey, Mouse (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Involucrin | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| involucrin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| It recognizes a protein of 66kDa-170kDa, identified as involucrin. In Western blotting of cultured human keratinocytes, this MAb reacts with a 120kDa protein. It stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∼115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig. Its epitope maps between codon 421-568 of human involucrin. Involucrin is expressed in a range of stratified squamous epithelia, including the cornea, which lacks a distinct cornified layer. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures, all cells that have left the basal layer express it. Involucrin expression is altered in pathological conditions: in psoriasis and other benign epidermal hyperplasias, involucrin expression begins closer to the basal layer than normal; expression is abnormal in squamous cell carcinomas and premalignant lesions, and is reduced in severe dysplasias of the larynx and cervix. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM259 | |
| Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P07476 | |
| IVL | |
| Purified involucrin from human keratinocytes | |
| 0.02 mg | |
| Extracellular Matrix | |
| 3713 | |
| Human, Pig, Canine, Primate, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction