Learn More
Iodic acid, 99%, Thermo Scientific Chemicals
Catalog Number p-7081777
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
7782-68-5
MFCD00011348
24345
CHEBI:24857
7782-68-5
MFCD00011348
24345
CHEBI:24857
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
TRUSTED_SUSTAINABILITY
Melting Point 110°C (decomposition)
Density 4.629
Odor Odorless
Sensitivity Light sensitive
Quantity 50 g
Molecular Formula HIO3
UN Number UN3085
Merck Index 14,5012
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
Solubility Information Soluble in water and methanol.
InChI Key ICIWUVCWSCSTAQ-UHFFFAOYSA-N
SMILES OI(=O)=O
IUPAC Name iodic acid
Molecular Weight (g/mol) 175.909
Formula Weight 175.91
Percent Purity 99%
Chemical Name or Material Iodic acid