Learn More
Iodine solution EU Pharmacopoeia 0.02M, Fisher Chemical™
Catalog Number 12920644
Click to view available options
Quantity:
1000 mL
Packaging:
Glass Bottle
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Concentration 0.02365M (0.0473N)
Packaging Glass Bottle
Quantity 1000 mL
Analyte or Component Names Iodine Solution