missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Ionomycin
Description
- Ionomycin is a potent and selective calcium ionophore derived from Streptomyces conglobatus (Liu et al.) It is used as a research tool to rapidly raise the intracellular level of calcium, and to study calcium transport across biological membranes by inducing the release of cytosolic calcium stores (Morgan & Jacob; Yoshida & Plant)
- Ionomycin is a more effective Ca++ ionophore than A23187, but less effective at binding and carrying Mg++ (Liu & Hermann)
- Ionomycin is able to activate and prime the polymorphonuclear neutrophil (PMN) oxidase (Elzi et al.), and is used in conjunction with Phorbol 12-myristate 13-acetate (PMA; Catalog #74042) for the activation of T cells (IC50 = 5.8 nM; Caraher et al.; Zhang et al.) This product is supplied as a 10 mg/mL solution in ethanol
- Activates T cells from human, mouse, or rat sources, in combination with PMA, to express cytokines including IL-17, IL-4, IL-10, and IL-2 (Caraher et al.; Harrington et al.; Parrish-Novak et al.)
Specifications
Specifications
| CAS | 56092-81-0; 64-17-5 |
| Synonym | SQ 23377 |
| Quantity | 1 mg |
| Molecular Formula | C41H72O9 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?