missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP3R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB120-5908-0.025ml
This item is not returnable.
View return policy
Description
IP3R1 Polyclonal specifically detects IP3R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| IP3R1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Q14643 | |
| ITPR1 | |
| Synthetic peptide corresponding to residues N(1829) K K K D D E V D R D A P S R K K A K E(1848) of human IP3 Receptor Type I. | |
| 0.025 mL | |
| Neuroscience | |
| 3708 | |
| Human | |
| Antisera |
| Western Blot | |
| Unconjugated | |
| Whole antisera diluted with PBS with 0.05% Sodium Azide | |
| DKFZp313E1334, DKFZp313N1434, inositol 14,5-triphosphate receptor, type 1, inositol 14,5-trisphosphate receptor type 1, INSP3R1, IP3 receptor, IP3 receptor isoform 1, IP3R, IP3R 1, IP3R1, SCA15, SCA16, spinocerebellar ataxia 15, spinocerebellar ataxia 16, Type 1 inositol 14,5-trisphosphate receptor, Type 1 InsP3 receptor | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| IP3R1 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction