missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP3R3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21400AF647
This item is not returnable.
View return policy
Description
IP3R3 Polyclonal antibody specifically detects IP3R3 in Human samples. It is validated for Western Blot
Specifications
| IP3R3 | |
| Polyclonal | |
| Western Blot | |
| FLJ36205, inositol 14,5-triphosphate receptor, type 3, inositol 14,5-trisphosphate receptor type 3, InsP3R3, IP3 receptor, IP3 receptor isoform 3, IP3R, IP3R 3, IP3R3, Type 3 inositol 14,5-trisphosphate receptor, Type 3 InsP3 receptor | |
| The immunogen recognized by this antibody maps to a region between residue 1755 and 1825 of human inositol 1,4,5-triphosphate receptor, type 3 using the numbering given in entry NP_002215.2 (GeneID 3710). | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 3710 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction