Learn More
Invitrogen™ IP6 Kinase Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570594
Product Code. 15849003
  • € 520.00 / 100µL
Description
IP6 Kinase Polyclonal Antibody for Western Blot
This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IP6 Kinase
Polyclonal
Unconjugated
PPIP5K1
diphosphoinositol pentakisphosphate kinase 1; HISPPD2A; histidine acid phosphatase domain containing 2A; histidine acid phosphatase domain-containing protein 2A; hsVIP1; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; inositol-hexakisphosphate kinase; insP6 and PP-IP5 kinase 1; IP6 kinase; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1; VIP1 homolog
Rabbit
Affinity chromatography
RUO
9677
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q6PFW1
PPIP5K1
synthetic peptide directed towards the N-terminal region of human PPIP5K1.