Learn More
Testo™ IP67 with TopSafe Waterproof Stainless Steel Food Probe (NTC)
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
Unit Size:
Each
Specifications
Specifications
| Type | Probe, Food (NTC) |
| Certifications/Compliance | EN 13485 and HACCP-compliant |
| Response Time | 8 sec. |
| Temperature Range (Metric) | -50°C to +150°C |