missing translation for 'onlineSavingsMsg'
Learn More

IP6K1 Antibody, Biotin, Novus Biologicals™

Product Code. 18645147 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18645147 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18645147 Supplier Novus Biologicals Supplier No. NBP318438B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6K1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 75 and 125..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9807
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.