missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP6K3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | IP6K3 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18483002
|
Novus Biologicals
NBP2-33689-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18113043
|
Novus Biologicals
NBP2-33689 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IP6K3 Polyclonal specifically detects IP6K3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IP6K3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q96PC2 | |
| 117283 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QRFYESLPLAMKRFTPQYKGTVTVHLWKDSTGHLSLVANPVKESQEPFKVSTESAAVAIWQTLQQTTGSNGSDCTLAQWPHAQL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| IHPK3ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, inositol hexakisphosphate kinase 3, Inositol hexaphosphate kinase 3EC 2.7.4.21, InsP6 kinase 3, INSP6K3MGC102928 | |
| IP6K3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title