Learn More
Tocris Bioscience™ IPA 3
Group I PAK inhibitor
€ 174.85 - € 670.64
Specifications
Purity | >96% |
---|---|
Product Type | IPA 3 |
Molecular Weight (g/mol) | 350.45 |
Inhibitors | IPA 3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18083832
|
Tocris Bioscience™
3622/10 |
10 mg |
€ 174.85
10mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18045902
|
Tocris Bioscience™
3622/50 |
50 mg |
€ 670.64
50mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Group I p21-activated kinase (PAK) inhibitor (IC50 = 2.5 μM at PAK1). Targets the autoregulatory mechanism and promotes the inactive conformation of PAKs. Inhibits PAK1-mediated signaling in vivo; potential anti-tumor agent.Negative Control also available.
Specifications
>96% | |
IPA 3 | |
350.45 | |
IPA 3 |
For Research Use Only