missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ipatasertib dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Ipatasertib dihydrochloride (GDC-0068 dihydrochloride) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Specifications
Specifications
| Chemical Name or Material | Ipatasertib dihydrochloride |
| Physical Form | Solid |
| CAS | 1396257-94-5 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H34Cl3N5O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | GDC-0068 dihydrochlorideRG-7440 dihydrochloride |
| Solubility Information | DMSO : 125 mg/mL (235.44 mM; Need ultrasonic) ∣H2O : ≥ 41 mg/mL (77.22 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction