Learn More
Ipatasertib, MedChemExpress
Product Code. 16422467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16422467
Brand: Medchem Express HY15186
MedChemExpress Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Specifications
Chemical Name or Material Ipatasertib
Physical Form Powder
CAS 1001264-89-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H32ClN5O2
For Use With (Application) Cancer-Kinase/protease
Synonym GDC-0068 RG7440
Solubility Information DMSO : 220 mg/mL (480.35 mM; Need ultrasonic)
SMILES ClC1=CC=C([C@@H](CNC(C)C)C(N2CCN(C3=C([C@H](C)C[C@H]4O)C4=NC=N3)CC2)=O)C=C1
Molecular Weight (g/mol) 458.0
Formula Weight 458.0
Percent Purity 95.0%
Grade Research
Show More Show Less
Research purposes only