missing translation for 'onlineSavingsMsg'
Learn More

IPCEF1 Antibody (OTI3G2), Biotin, Novus Biologicals™

Product Code. 18191953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18191953 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18191953 Supplier Novus Biologicals Supplier No. NBP271781B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IPCEF1 Monoclonal specifically detects IPCEF1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPCEF1
Applications Western Blot
Classification Monoclonal
Clone OTI3G2
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias interaction protein for cytohesin exchange factors 1, interactor protein for cytohesin exchange factors 1, KIAA0403interaction protein for cytohesin exchange factors 1 Interaction protein forcytohesin exchange factors 1, phosphoinositide binding protein PIP3-E, Phosphoinositide-binding protein PIP3-E, PIP3-E, RP3-402L9.2
Gene Symbols IPCEF1
Host Species Mouse
Immunogen Full length recombinant protein of human IPCEF1 (NP_056368) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26034.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.