Learn More
Iproniazid, TRC
Specifications
Specifications
| Chemical Name or Material | Iproniazid |
| CAS | 54-92-2 |
| Molecular Formula | C9 H13 N3 O |
| Quantity | 500 mg |
| Synonym | 4-Pyridinecarboxylic acid, 2-(1-methylethyl)hydrazide, Isonicotinic acid, 2-isopropylhydrazide (6CI,8CI), 1-Isonicotinoyl-2-isopropylhydrazine, Euphozid, IPN, Iprazid, Iprazide, Iproniazid, Iproniazide, Isonicotinic acid N'-isopropyl hydrazide, Marsalid, Marsilid, N'-Isopropylisonicotinoylhydrazide |
| InChI Formula | InChI=1S/C9H13N3O/c1-7(2)11-12-9(13)8-3-5-10-6-4-8/h3-7,11H,1-2H3,(H,12,13) |
| SMILES | CC(C)NNC(=O)c1ccncc1 |
| IUPAC Name | N'-propan-2-ylpyridine-4-carbohydrazide |
| Molecular Weight (g/mol) | 179.22 |
| Formula Weight | 179.1059 |