Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Hcp50
Product Code. 16324877 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
110, 260, 750 Models Constant Climate Chamber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16324877 110, 260, 750 Models Constant Climate Chamber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16324877 Supplier Memmert™ Supplier No. HCP50D00136
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Works test data, temperature distribution survey
Includes temperature distribution survey for one free-selectable temperature and humidity value.
  • IQ/OQ document with works test data for chamber
  • 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP and ICH)
Specifications
For Use With (Equipment) 110, 260, 750 Models Constant Climate Chamber
Product Type IQ/OQ Documentation
For Use With (Application) One Free-Selectable Temperature and Humidity Value for 27 Measuring Points to DIN 12880:2007-05