missing translation for 'onlineSavingsMsg'
Learn More

IQCB1 Antibody - BSA Free, Novus Biologicals™

Product Code. 30559375 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30559375 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30559375 Supplier Novus Biologicals Supplier No. NBP347313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IQCB1 Polyclonal antibody specifically detects IQCB1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCB1
Applications ELISA, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias IQ calmodulin-binding motif containing 1, IQ motif containing B1, KIAA0036nephrocystin 5, Nephrocystin-5, NPHP5IQ calmodulin-binding motif-containing protein 1, p53 and DNA damage-regulated IQ motif protein, PIQ, SLSN5
Host Species Rabbit
Immunogen IQ motif containing B1. (Uniprot# Q15051)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9657
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.