missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IQCB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29386
This item is not returnable.
View return policy
Description
IQCB1 Polyclonal antibody specifically detects IQCB1 in Human samples. It is validated for Immunoprecipitation
Specifications
| IQCB1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9657 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| IQ calmodulin-binding motif containing 1, IQ motif containing B1, KIAA0036nephrocystin 5, Nephrocystin-5, NPHP5IQ calmodulin-binding motif-containing protein 1, p53 and DNA damage-regulated IQ motif protein, PIQ, SLSN5 | |
| Between 548 and 598 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction