missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQGAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 127.00 - € 387.00
Specifications
| Antigen | IQGAP1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649853
|
Novus Biologicals
NBP3-21443-25ul |
25 μg |
€ 127.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600615
|
Novus Biologicals
NBP3-21443-100ul |
100 μg |
€ 387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IQGAP1 Polyclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IQGAP1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.0 | |
| 8826 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195RasGAP-like with IQ motifs, SAR1 | |
| Produced in rabbits immunized with E. coli-derived Human IQGAP1 fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title