Learn More
IQGAP1 Monoclonal Antibody (OTI6E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809921S
This item is not returnable.
View return policy
Description
IQGAP1 Monoclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IQGAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P37837 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E1 | |
| Unconjugated | |
| TALDO1 | |
| AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 | |
| Full length human recombinant protein of human TALDO1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |