missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519189
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
Specifications
| IQGAP1 | |
| Polyclonal | |
| Unconjugated | |
| IQGAP1 | |
| AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P46940 | |
| IQGAP1 | |
| Synthetic peptide sequence (KRQSGQTDPLQKEE) corresponding to the internal amino acids of IQGAP1 (aa 357-371). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction