missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP2 Recombinant Rabbit Monoclonal Antibody (23GB4020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | IQGAP2 |
|---|---|
| Clone | 23GB4020 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089657
![]() |
Invitrogen™
MA553672 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087724
|
Invitrogen™
MA553673 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IQGAP2 Recombinant Monoclonal antibody specifically detects IQGAP2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| IQGAP2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13576 | |
| 10788 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4020 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 | |
| A synthesized peptide derived from human IQGAP2 (1301-1495AA). | |
| Primary | |
| -20°C | |
| IQGAP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title