Learn More
Unity Lab Services™ IQOQ Incubators, Ovens, Vacuum Oven And Furnaces 3rd Party
Description
- Installation and operational qualification service of ovens, vacuum ovens, furnaces and incubators manufactured by third parties.
- This service will confirm equipment is operating to defined specifications.
- Operational qualification includes a test of temperature distribution with 5 points on 1 shelf for vacuum oven and 9 points in the work volume for ovens and incubators.
- The service includes a test of temperature distribution at one temperature setpoint.
- NOTE! For the support of Memmert products, validation products must be ordered separately under item code 16344567
Specifications
Specifications
| For Use With (Equipment) | Ovens, Vacuums, Furnaces, Incubators |
| Product Type | IQ/OQ Service |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?