Learn More
Grant Bio™ IQ/OQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Merk:  Grant Bio™ IQOQ MSH-300i
Overige gegevens : Gewicht : 0.01000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15400494
  • € 485.52 / 1 stuk
Geschatte verzenddatum
bij leverancier 28-02-2018
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
  • Operational Qualification (OQ): instrument is functioning in accordance with its specification
  • Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
Specificatie
Specificatie
IQ/OQ Documentation
Grant MSH-300i Digital Magnetic Stirring Hotplate
Video's